Assay Details
| Gene Symbol: HUB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198304.1 | NP_001185233.1 | 4 - 5 | 779 | 134 | |
| NM_001333696.1 | NP_001321104.1 | 4 - 5 | 856 | 134 | ||
| NM_104398.4 | NP_564680.4 | 4 - 5 | 652 | 134 | ||
More Information
Set Membership: |
|

HUB2
Genomic Map