Assay Details
| Gene Symbol: HUB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160956.1 | NP_001154428.1 | 1 - 2 | 125 | 88 | |
| NM_001198304.1 | NP_001185233.1 | 12 - 13 | 1768 | 88 | ||
| NM_001198305.2 | NP_001185234.1 | 1 - 2 | 569 | 88 | ||
| NM_001333696.1 | NP_001321104.1 | 11 - 12 | 1833 | 88 | ||
| NM_104398.4 | NP_564680.4 | 11 - 12 | 1629 | 88 | ||
| GenBank mRNA | BT029207.1 | - | 1 - 2 | 78 | 88 | |
More Information
Set Membership: |
|

HUB2
Genomic Map