Assay Details
| Gene Symbol: HUB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160956.1 | NP_001154428.1 | 7 - 8 | 901 | 98 | |
| NM_001198304.1 | NP_001185233.1 | 18 - 19 | 2544 | 98 | ||
| NM_001198305.2 | NP_001185234.1 | 7 - 8 | 1345 | 98 | ||
| NM_001333696.1 | NP_001321104.1 | 17 - 18 | 2609 | 98 | ||
| NM_104398.4 | NP_564680.4 | 17 - 18 | 2405 | 98 | ||
| GenBank mRNA | BT029207.1 | - | 7 - 8 | 854 | 98 | |
More Information
Set Membership: |
|

HUB2
Genomic Map