Assay Details
| Gene Symbol: HAC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333753.1 | NP_001319247.1 | 7 - 8 | 2820 | 119 | |
| NM_104475.3 | NP_564706.1 | 7 - 8 | 2882 | 119 | ||
More Information
Set Membership: |
|

HAC4
Genomic Map