Assay Details
| Gene Symbol: EMB3108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333874.1 | NP_001321198.1 | 1 - 2 | 501 | 76 | |
| NM_104691.4 | NP_176207.1 | 1 - 2 | 826 | 76 | ||
| GenBank mRNA | AF424570.1 | - | 1 - 2 | 430 | 76 | |
| AY142022.1 | - | 1 - 2 | 326 | 76 | ||
More Information
Set Membership: |
|

EMB3108
Genomic Map