Assay Details
| Gene Symbol: BGLU4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333876.1 | NP_001322340.1 | 12 - 13 | 1298 | 134 | |
| NM_104701.3 | NP_176217.2 | 12 - 13 | 1296 | 134 | ||
More Information
Set Membership: |
|

BGLU4
Genomic Map