Assay Details
| Gene Symbol: SC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198349.1 | NP_001185278.1 | 10 - 11 | 911 | 98 | |
| NM_104806.4 | NP_176320.1 | 10 - 11 | 997 | 98 | ||
| GenBank mRNA | AF225920.1 | - | 10 - 11 | 777 | 98 | |
| AY045788.1 | - | 10 - 11 | 840 | 98 | ||
| AY079336.1 | - | 10 - 11 | 714 | 98 | ||
| AY086742.1 | - | 10 - 11 | 829 | 98 | ||
More Information
Set Membership: |
|

SC3
Genomic Map