Assay Details
| Gene Symbol: SC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198349.1 | NP_001185278.1 | 1 - 2 | 250 | 64 | |
| NM_104806.4 | NP_176320.1 | 1 - 2 | 336 | 64 | ||
| GenBank mRNA | AF225920.1 | - | 1 - 2 | 116 | 64 | |
| AY045788.1 | - | 1 - 2 | 179 | 64 | ||
| AY079336.1 | - | 1 - 2 | 53 | 64 | ||
| AY086742.1 | - | 1 - 2 | 168 | 64 | ||
More Information
Set Membership: |
|

SC3
Genomic Map