Assay Details
| Gene Symbol: SC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198349.1 | NP_001185278.1 | 4 - 5 | 465 | 77 | |
| NM_104806.4 | NP_176320.1 | 4 - 5 | 551 | 77 | ||
| GenBank mRNA | AF225920.1 | - | 4 - 5 | 331 | 77 | |
| AY045788.1 | - | 4 - 5 | 394 | 77 | ||
| AY079336.1 | - | 4 - 5 | 268 | 77 | ||
| AY086742.1 | - | 4 - 5 | 383 | 77 | ||
More Information
Set Membership: |
|

SC3
Genomic Map