Assay Details
| Gene Symbol: BGLU45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198358.1 | NP_001185287.1 | 5 - 6 | 454 | 167 | |
| NM_001334002.1 | NP_001319288.1 | 5 - 6 | 548 | 167 | ||
| NM_001334003.1 | NP_001322277.1 | 5 - 6 | 452 | 167 | ||
| NM_001334004.1 | NP_001322278.1 | 2 - 3 | 245 | 167 | ||
| NM_104863.3 | NP_176374.1 | 5 - 6 | 548 | 167 | ||
More Information
Set Membership: |
|

BGLU45
Genomic Map