Assay Details
| Gene Symbol: BGLU45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198358.1 | NP_001185287.1 | 8 - 9 | 916 | 114 | |
| NM_001334003.1 | NP_001322277.1 | 8 - 9 | 914 | 114 | ||
| NM_001334004.1 | NP_001322278.1 | 5 - 6 | 707 | 114 | ||
| NM_104863.3 | NP_176374.1 | 8 - 9 | 1010 | 114 | ||
More Information
Set Membership: |
|

BGLU45
Genomic Map