Assay Details
| Gene Symbol: AT2G34590 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129013.3 | NP_181006.1 | 1 - 2 | 407 | 72 | |
| GenBank mRNA | AF167983.1 | - | 1 - 2 | 236 | 72 | |
| BT025968.1 | - | 1 - 2 | 108 | 72 | ||
More Information
Set Membership: |
|

AT2G34590
Genomic Map