Assay Details
| Gene Symbol: JAZ7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129014.4 | NP_181007.1 | 1 - 2 | 267 | 83 | |
| GenBank mRNA | BT010666.1 | - | 1 - 2 | 217 | 83 | |
| BT010963.1 | - | 1 - 2 | 125 | 83 | ||
More Information
Set Membership: |
|

JAZ7
Genomic Map