Assay Details
| Gene Symbol: ACD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129023.5 | NP_181016.1 | 3 - 4 | 959 | 78 | |
| GenBank mRNA | BT012177.1 | - | 3 - 4 | 520 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACD11| Gene Symbol: ACD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129023.5 | NP_181016.1 | 3 - 4 | 959 | 78 | |
| GenBank mRNA | BT012177.1 | - | 3 - 4 | 520 | 78 | |
Set Membership: |
|