Assay Details
| Gene Symbol: RBOH F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105079.3 | NP_564821.1 | 8 - 9 | 2503 | 146 | |
| GenBank mRNA | AF015301.1 | - | 8 - 9 | 2502 | 146 | |
More Information
Set Membership: |
|

RBOH F
Genomic Map