Assay Details
| Gene Symbol: RBOH F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105079.3 | NP_564821.1 | 9 - 10 | 2670 | 62 | |
| GenBank mRNA | AF015301.1 | - | 9 - 10 | 2669 | 62 | |
More Information
Set Membership: |
|

RBOH F
Genomic Map