Assay Details
| Gene Symbol: AGL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105204.3 | NP_176709.2 | 1 | 454 | 155 | |
| GenBank mRNA | AY141245.1 | - | 1 | 454 | 155 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL38| Gene Symbol: AGL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105204.3 | NP_176709.2 | 1 | 454 | 155 | |
| GenBank mRNA | AY141245.1 | - | 1 | 454 | 155 | |
Set Membership: |
|