Assay Details
| Gene Symbol: UNE15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340867.1 | NP_001329208.1 | 3 - 4 | 2515 | 59 | |
| NM_117430.3 | NP_193092.1 | 2 - 3 | 3862 | 59 | ||
| GenBank mRNA | AK118342.1 | - | 2 - 3 | 319 | 59 | |
| BT004928.1 | - | 2 - 3 | 247 | 59 | ||
More Information
Set Membership: |
|

UNE15
Genomic Map