Assay Details
| Gene Symbol: MEE57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117435.1 | NP_193097.1 | 4 - 5 | 2612 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MEE57| Gene Symbol: MEE57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117435.1 | NP_193097.1 | 4 - 5 | 2612 | 140 | |
Set Membership: |
|