Assay Details
| Gene Symbol: GR-RBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084916.1 | NP_001078385.1 | 2 - 3 | 193 | 81 | |
| NM_001084917.1 | NP_001078386.1 | 2 - 3 | 193 | 81 | ||
| NM_117459.4 | NP_193121.1 | 2 - 3 | 431 | 81 | ||
| NM_179046.2 | NP_849377.1 | 2 - 3 | 193 | 81 | ||
| GenBank mRNA | AY072361.1 | - | 2 - 3 | 189 | 81 | |
| AY085621.1 | - | 2 - 3 | 192 | 81 | ||
| BT002197.1 | - | 1 - 2 | 123 | 81 | ||
More Information
Set Membership: |
|

GR-RBP2
Genomic Map