Assay Details
| Gene Symbol: ORC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117553.2 | NP_567440.1 | 1 | 275 | 145 | |
| GenBank mRNA | AJ421410.1 | - | 1 | 270 | 145 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ORC1A| Gene Symbol: ORC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117553.2 | NP_567440.1 | 1 | 275 | 145 | |
| GenBank mRNA | AJ421410.1 | - | 1 | 270 | 145 | |
Set Membership: |
|