Assay Details
| Gene Symbol: ABCG30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341010.1 | NP_001319944.1 | 4 - 5 | 1125 | 100 | |
| NM_001341011.1 | NP_001328800.1 | 4 - 5 | 1131 | 100 | ||
| NM_001341012.1 | NP_001328801.1 | 5 - 6 | 831 | 100 | ||
| NM_117611.5 | NP_193258.3 | 4 - 5 | 1131 | 100 | ||
More Information
Set Membership: |
|

ABCG30
Genomic Map