Assay Details
| Gene Symbol: CBL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341098.1 | NP_001328805.1 | 6 - 7 | 602 | 108 | |
| NM_117730.3 | NP_567492.1 | 7 - 8 | 666 | 108 | ||
| GenBank mRNA | AF192884.1 | - | 6 - 7 | 602 | 108 | |
| AK176189.1 | - | 7 - 8 | 666 | 108 | ||
More Information
Set Membership: |
|

CBL6
Genomic Map