Assay Details
| Gene Symbol: BIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341302.1 | NP_001319987.1 | 4 - 5 | 1085 | 89 | |
| NM_117987.4 | NP_193606.1 | 4 - 5 | 850 | 89 | ||
| GenBank mRNA | AY086529.1 | - | 4 - 5 | 686 | 89 | |
| BT026031.1 | - | 3 - 4 | 514 | 89 | ||
More Information
Set Membership: |
|

BIN2
Genomic Map