Assay Details
| Gene Symbol: DIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334269.1 | NP_001319330.1 | 4 - 5 | 1218 | 82 | |
| NM_105377.5 | NP_176878.1 | 3 - 4 | 1491 | 82 | ||
| GenBank mRNA | AK176481.1 | - | 3 - 4 | 1054 | 82 | |
| BT025995.1 | - | 3 - 4 | 1011 | 82 | ||
More Information
Set Membership: |
|

DIN9
Genomic Map