Assay Details
| Gene Symbol: AT1G67640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105432.2 | NP_176932.1 | 4 - 5 | 1281 | 139 | |
| GenBank mRNA | DQ446407.1 | - | 4 - 5 | 1163 | 139 | |
More Information
Set Membership: |
|

AT1G67640
Genomic Map