Assay Details
| Gene Symbol: FKF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105475.4 | NP_564919.1 | 1 - 2 | 371 | 83 | |
| GenBank mRNA | AY064999.1 | - | 1 - 2 | 335 | 83 | |
| AY113029.1 | - | 1 - 2 | 273 | 83 | ||
More Information
Set Membership: |
|

FKF1
Genomic Map