Assay Details
| Gene Symbol: IPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105517.2 | NP_177013.1 | 1 | 1314 | 74 | |
| GenBank mRNA | AB062607.1 | - | 1 | 1018 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IPT1| Gene Symbol: IPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105517.2 | NP_177013.1 | 1 | 1314 | 74 | |
| GenBank mRNA | AB062607.1 | - | 1 | 1018 | 74 | |
Set Membership: |
|