Assay Details
| Gene Symbol: LPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105773.3 | NP_565008.1 | 2 - 3 | 1621 | 88 | |
| GenBank mRNA | AY050818.1 | - | 2 - 3 | 1460 | 88 | |
| AY091420.1 | - | 2 - 3 | 1362 | 88 | ||
More Information
Set Membership: |
|

LPR2
Genomic Map