Assay Details
| Gene Symbol: GER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105920.2 | NP_177405.1 | 1 | 859 | 98 | |
| GenBank mRNA | U75197.1 | - | 1 | 789 | 98 | |
| U75201.1 | - | 1 | 796 | 98 | ||
| U75206.1 | - | 1 | 796 | 98 | ||
More Information
Set Membership: |
|

GER1
Genomic Map