Assay Details
| Gene Symbol: AT1G73240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105983.3 | NP_565060.1 | 3 - 4 | 1348 | 93 | |
| GenBank mRNA | AY087848.1 | - | 3 - 4 | 1157 | 93 | |
More Information
Set Membership: |
|

AT1G73240
Genomic Map