Assay Details
| Gene Symbol: GSR 1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123119.4 | NP_198576.1 | 4 - 5 | 499 | 108 | |
| GenBank mRNA | AF419608.1 | - | 4 - 5 | 343 | 108 | |
| AF428386.1 | - | 4 - 5 | 343 | 108 | ||
| AY079113.1 | - | 4 - 5 | 270 | 108 | ||
| BT000753.1 | - | 4 - 5 | 350 | 108 | ||
More Information
Set Membership: |
|

GSR 1
Genomic Map