Assay Details
| Gene Symbol: EMB2656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123122.4 | NP_198579.2 | 5 - 6 | 1333 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMB2656| Gene Symbol: EMB2656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123122.4 | NP_198579.2 | 5 - 6 | 1333 | 60 | |
Set Membership: |
|