Assay Details
| Gene Symbol: NF-YC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123174.5 | NP_198630.2 | 7 - 8 | 970 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NF-YC12| Gene Symbol: NF-YC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123174.5 | NP_198630.2 | 7 - 8 | 970 | 103 | |
Set Membership: |
|