Assay Details
| Gene Symbol: AT3G42950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114163.4 | NP_189881.1 | 4 - 5 | 1229 | 71 | |
| GenBank mRNA | AY050383.1 | - | 4 - 5 | 1117 | 71 | |
More Information
Set Membership: |
|

AT3G42950
Genomic Map