Assay Details
| Gene Symbol: DPBF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114314.5 | NP_566870.1 | 2 - 3 | 899 | 92 | |
| GenBank mRNA | AJ419600.1 | - | 2 - 3 | 840 | 92 | |
| BT002923.1 | - | 2 - 3 | 899 | 92 | ||
| BT004342.1 | - | 1 - 2 | 802 | 92 | ||
More Information
Set Membership: |
|

DPBF2
Genomic Map