Assay Details
| Gene Symbol: HDA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114336.4 | NP_190054.2 | 3 - 4 | 527 | 95 | |
| GenBank mRNA | BT002003.1 | - | 3 - 4 | 437 | 95 | |
| BT006576.1 | - | 3 - 4 | 270 | 95 | ||
More Information
Set Membership: |
|

HDA9
Genomic Map