Assay Details
| Gene Symbol: HDA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_114336.4 | NP_190054.2 | 9 - 10 | 1135 | 131 | |
| GenBank mRNA | BT002003.1 | - | 9 - 10 | 1045 | 131 | |
| BT006576.1 | - | 9 - 10 | 878 | 131 | ||
More Information
Set Membership: |
|

HDA9
Genomic Map