Assay Details
| Gene Symbol: HDA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203083.1 | NP_001190012.1 | 2 - 3 | 153 | 93 | |
| NM_114344.4 | NP_566872.1 | 2 - 3 | 476 | 93 | ||
| GenBank mRNA | AF195545.1 | - | 2 - 3 | 135 | 93 | |
| AY065396.1 | - | 2 - 3 | 149 | 93 | ||
| AY096729.1 | - | 1 - 2 | 87 | 93 | ||
More Information
Set Membership: |
|

HDA3
Genomic Map