Assay Details
| Gene Symbol: HDA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203083.1 | NP_001190012.1 | 5 - 6 | 494 | 75 | |
| NM_114344.4 | NP_566872.1 | 5 - 6 | 826 | 75 | ||
| GenBank mRNA | AF195545.1 | - | 5 - 6 | 485 | 75 | |
| AY065396.1 | - | 5 - 6 | 499 | 75 | ||
| AY096729.1 | - | 4 - 5 | 437 | 75 | ||
More Information
Set Membership: |
|

HDA3
Genomic Map