Assay Details
| Gene Symbol: HEN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126675.3 | NP_565338.1 | 15 - 16 | 3110 | 144 | |
| GenBank mRNA | BT005794.1 | - | 15 - 16 | 2879 | 144 | |
More Information
Set Membership: |
|

HEN2
Genomic Map