Assay Details
| Gene Symbol: NAD-ME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126936.4 | NP_178980.1 | 17 - 18 | 1849 | 88 | |
| GenBank mRNA | AY091108.1 | - | 17 - 18 | 1814 | 88 | |
| BT000996.1 | - | 17 - 18 | 1688 | 88 | ||
More Information
Set Membership: |
|

NAD-ME1
Genomic Map