Assay Details
| Gene Symbol: TIF3H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100960.3 | NP_563880.1 | 11 - 12 | 1273 | 78 | |
| NM_202079.1 | NP_973808.1 | 10 - 11 | 1159 | 78 | ||
| GenBank mRNA | AY054641.1 | - | 11 - 12 | 1107 | 78 | |
| AY081546.1 | - | 10 - 11 | 956 | 78 | ||
| AY087338.1 | - | 11 - 12 | 1175 | 78 | ||
| BT000765.1 | - | 11 - 12 | 1052 | 78 | ||
More Information
Set Membership: |
|

TIF3H1
Genomic Map