Assay Details
| Gene Symbol: AAPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332070.1 | NP_001323307.1 | 9 - 10 | 598 | 141 | |
| NM_101226.5 | NP_172813.1 | 11 - 12 | 967 | 141 | ||
| NM_202088.2 | NP_973817.1 | 10 - 11 | 864 | 141 | ||
| GenBank mRNA | AF091843.1 | - | 11 - 12 | 804 | 141 | |
| AY080725.1 | - | 11 - 12 | 841 | 141 | ||
| AY114062.1 | - | 10 - 11 | 705 | 141 | ||
More Information
Set Membership: |
|

AAPT1
Genomic Map