Assay Details
| Gene Symbol: AAPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332070.1 | NP_001323307.1 | 12 - 13 | 789 | 75 | |
| NM_101226.5 | NP_172813.1 | 14 - 15 | 1158 | 75 | ||
| NM_202088.2 | NP_973817.1 | 13 - 14 | 1055 | 75 | ||
| GenBank mRNA | AF091843.1 | - | 14 - 15 | 995 | 75 | |
| AY080725.1 | - | 14 - 15 | 1032 | 75 | ||
| AY114062.1 | - | 13 - 14 | 896 | 75 | ||
More Information
Set Membership: |
|

AAPT1
Genomic Map