Assay Details
| Gene Symbol: AAPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332070.1 | NP_001323307.1 | 14 - 15 | 912 | 82 | |
| NM_101226.5 | NP_172813.1 | 16 - 17 | 1281 | 82 | ||
| NM_202088.2 | NP_973817.1 | 15 - 16 | 1178 | 82 | ||
| GenBank mRNA | AF091843.1 | - | 16 - 17 | 1118 | 82 | |
| AY080725.1 | - | 16 - 17 | 1155 | 82 | ||
| AY114062.1 | - | 15 - 16 | 1019 | 82 | ||
More Information
Set Membership: |
|

AAPT1
Genomic Map