Assay Details
| Gene Symbol: AAPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332070.1 | NP_001323307.1 | 16 - 17 | 1045 | 76 | |
| NM_101226.5 | NP_172813.1 | 18 - 19 | 1414 | 76 | ||
| NM_202088.2 | NP_973817.1 | 17 - 18 | 1311 | 76 | ||
| GenBank mRNA | AF091843.1 | - | 18 - 19 | 1251 | 76 | |
| AY080725.1 | - | 18 - 19 | 1288 | 76 | ||
| AY114062.1 | - | 17 - 18 | 1152 | 76 | ||
More Information
Set Membership: |
|

AAPT1
Genomic Map