Assay Details
| Gene Symbol: SUVR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337514.1 | NP_001327806.1 | 2 - 3 | 1160 | 67 | |
| NM_001337515.1 | NP_001327808.1 | 2 - 3 | 1126 | 67 | ||
| NM_001337516.1 | NP_001327807.1 | 3 - 4 | 1061 | 67 | ||
| NM_001337517.1 | NP_001327809.1 | 3 - 4 | 1045 | 67 | ||
| NM_111309.3 | NP_187088.2 | 2 - 3 | 1147 | 67 | ||
| NM_202488.2 | NP_974217.1 | 3 - 4 | 1066 | 67 | ||
More Information
Set Membership: |
|

SUVR4
Genomic Map