Assay Details
| Gene Symbol: SUVR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337514.1 | NP_001327806.1 | 7 - 8 | 1702 | 128 | |
| NM_111309.3 | NP_187088.2 | 7 - 8 | 1689 | 128 | ||
| NM_202488.2 | NP_974217.1 | 8 - 9 | 1608 | 128 | ||
More Information
Set Membership: |
|

SUVR4
Genomic Map