Assay Details
| Gene Symbol: SWP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111346.4 | NP_187125.1 | 5 - 6 | 2507 | 73 | |
| GenBank mRNA | AF466359.1 | - | 5 - 6 | 2569 | 73 | |
| AJ511271.1 | - | 5 - 6 | 2569 | 73 | ||
More Information
Set Membership: |
|

SWP
Genomic Map